missing translation for 'onlineSavingsMsg'
Learn More

Poeppelmann™ GPN 600 Series Natural LDPE Tapered Closure

€ 12.55 - € 231.00

Specifications

Color Natural
Material LDPE
Type Tapered Closure
For Use With (Application) For use as plugs in bores or internal thread, or as caps for external thread
View More Specs

Products 12
Product Code Brand Diameter (Metric) Outside Length (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Outside Length (Metric) Price Quantity & Availability  
10175204
View Documents
Poeppelmann™
60001740000
21 mm 8.8 mm
€ 12.55
Pack of 100
Estimated Shipment: 13-03-2025
to see stock.
Add to basket
 
10626845
View Documents
Poeppelmann™
60011430000
124.5 mm 28.7 mm
€ 114.00
Pack of 50
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
 
10421595
View Documents
Poeppelmann™
60001820000
22 mm 8.8 mm
€ 16.25
Pack of 100
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
 
10665885
View Documents
Poeppelmann™
60023120000
249.5 mm 33.5 mm
€ 214.00
Pack of 25
Estimated Shipment: 18-02-2025
to see stock.
Add to basket
 
10368574
View Documents
Poeppelmann™
60025300000
265 mm 18 mm
€ 231.00
Pack of 25
In Stock
to see stock.
Edge
Add to basket
 
10358974
View Documents
Poeppelmann™
60029340000
314 mm 33 mm
€ 230.00
Pack of 25
Estimated Shipment: 18-02-2025
to see stock.
Add to basket
 
10678365
View Documents
Poeppelmann™
60002570000
30 mm 13.8 mm
€ 16.75
Pack of 100
Estimated Shipment: 12-02-2025
to see stock.
Add to basket
 
10286974
View Documents
Poeppelmann™
60001870000
24 mm 8.7 mm
€ 14.00
Pack of 100
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
 
10398764
View Documents
Poeppelmann™
60003790000
44 mm 13.8 mm
€ 26.95
Pack of 100
Estimated Shipment: 18-02-2025
to see stock.
Add to basket
 
10657415
View Documents
Poeppelmann™
60005700000
63 mm 13.5 mm
€ 63.80
Pack of 100
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
 
10083434
View Documents
Poeppelmann™
60007650000
87 mm 17 mm
€ 155.00
Pack of 100
Estimated Shipment: 17-02-2025
to see stock.
Add to basket
 
10718405
View Documents
Poeppelmann™
60006250000
68 mm 13.5 mm
€ 92.10
Pack of 100
Estimated Shipment: 18-02-2025
to see stock.
Add to basket
 
Description

Description

Specifications

Specifications

Natural
LDPE
Tapered Closure
For use as plugs in bores or internal thread, or as caps for external thread
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.