Learn More
Poeppelmann™ GPN 600 Series Natural LDPE Tapered Closure
Brand:  Poeppelmann™ 60001870000
 View more versions of this product
Product Code. 10286974
  • € 14.00 / Pack of 100
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Specifications
Natural
8.7 mm
O.D. x H: 24 x 8.7 mm, 8.2 mm Insertion Depth
LDPE
For use as plugs in bores or internal thread, or as caps for external thread
100 Pack
24 mm
Tapered Closure