Learn More
Poeppelmann™ GPN 600 Series Natural LDPE Tapered Closure
Brand: Poeppelmann™ 60025300000
Specifications
Natural | |
18 mm | |
O.D. x H: 265 x 18 mm, 18 mm Insertion Depth | |
LDPE |
For use as plugs in bores or internal thread, or as caps for external thread | |
25 Pack | |
265 mm | |
Tapered Closure |