Learn More
Poeppelmann™ GPN 600 Series Natural LDPE Tapered Closure
Brand: Poeppelmann™ 60001740000
This item is not returnable.
View return policy
Specifications
Natural | |
8.8 mm | |
O.D. x H: 21 x 8.8 mm, 9.2 mm Insertion Depth | |
LDPE |
For use as plugs in bores or internal thread, or as caps for external thread | |
100 Pack | |
21 mm | |
Tapered Closure |