Learn More
Poeppelmann™ GPN 600 Series Natural LDPE Tapered Closure
Brand: Poeppelmann™ 60005700000
This item is not returnable.
View return policy
Specifications
Natural | |
13.5 mm | |
O.D. x H: 63 x 13.5 mm, 13.4 mm Insertion Depth | |
LDPE |
For use as plugs in bores or internal thread, or as caps for external thread | |
100 Pack | |
63 mm | |
Tapered Closure |