Learn More
Poeppelmann™ GPN 600 Series Natural LDPE Tapered Closure
Brand:  Poeppelmann™ 60011430000
 View more versions of this product
Product Code. 10626845
  • € 114.00 / Pack of 50
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Natural
28.7 mm
O.D. x H: 124.5 x 28.7 mm, 28 mm Insertion Depth
LDPE
For use as plugs in bores or internal thread, or as caps for external thread
50 Pack
124.5 mm
Tapered Closure
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers