Learn More
Poeppelmann™ GPN 600 Series Natural LDPE Tapered Closure
Brand:  Poeppelmann™ 60005700000
 View more versions of this product
Product Code. 10657415
  • € 63.80 / Pack of 100
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Specifications
Natural
13.5 mm
O.D. x H: 63 x 13.5 mm, 13.4 mm Insertion Depth
LDPE
For use as plugs in bores or internal thread, or as caps for external thread
100 Pack
63 mm
Tapered Closure