Learn More
Poeppelmann™ GPN 600 Series Natural LDPE Tapered Closure
Brand:  Poeppelmann™ 60001740000
 View more versions of this product
Product Code. 10175204
  • € 12.55 / Pack of 100
Estimated Shipment: 13-03-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Natural
8.8 mm
O.D. x H: 21 x 8.8 mm, 9.2 mm Insertion Depth
LDPE
For use as plugs in bores or internal thread, or as caps for external thread
100 Pack
21 mm
Tapered Closure
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers