Learn More
Poeppelmann™ GPN 600 Series Natural LDPE Tapered Closure
Brand:  Poeppelmann™ 60029340000
 View more versions of this product
Product Code. 10358974
  • € 230.00 / Pack of 25
Estimated Shipment: 07-01-2025
to see stock.
Explore more special offers
Specifications
Natural
33 mm
O.D. x H: 314 x 33 mm, 32.5 mm Insertion Depth
LDPE
For use as plugs in bores or internal thread, or as caps for external thread
25 Pack
314 mm
Tapered Closure