Learn More
Poeppelmann™ KAPSTO™ GPN 320 Finned Plugs
Featuring ribs on the shake to provide a secure fit even with varying tube tolerances. Poeppelmann™ KAPSTO™ GPN 320 Finned Plugs are manufactured from black PE-LLD plastic.
€ 20.55 - € 133.00
Specifications
Product Type | Ribbed Closure Plug |
---|---|
Certifications/Compliance | DIN EN 10220, DIN EN 10255 and DIN EN ISO 1127th |
For Use With (Application) | For high loading capacity |
Product Code | Brand | Diameter (Metric) Outer | Height (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Height (Metric) | Price | Quantity & Availability | |||||
10615885
|
Poeppelmann™
32001020000 |
102 mm | 22 mm |
€ 133.00
Pack of 25 |
Estimated Shipment: 12-05-2025
Log in to see stock. |
|||||
10044014
|
Poeppelmann™
32000180000 |
18 mm | 15 mm |
€ 20.55
Pack of 100 |
Estimated Shipment: 21-05-2025
Log in to see stock. |
|||||
10298284
|
Poeppelmann™
32000200000 |
20 mm | 15 mm |
€ 23.70
Pack of 100 |
Estimated Shipment: 23-06-2025
Log in to see stock. |
|||||
10707605
|
Poeppelmann™
32000320000 |
32 mm | 19 mm |
€ 58.55
Pack of 100 |
Estimated Shipment: 03-06-2025
Log in to see stock. |
|||||
10388654
|
Poeppelmann™
32000340000 |
34 mm | 19 mm |
€ 65.25
Pack of 100 |
Estimated Shipment: 03-06-2025
Log in to see stock. |
|||||
10398654
|
Poeppelmann™
32000380000 |
38 mm | 19 mm |
€ 82.80
Pack of 100 |
Estimated Shipment: 03-06-2025
Log in to see stock. |
|||||
10490045
|
Poeppelmann™
32000400000 |
40 mm | 19 mm |
€ 103.00
Pack of 100 |
Estimated Shipment: 01-07-2025
Log in to see stock. |
|||||
10748175
|
Poeppelmann™
32000480000 |
48 mm | 19 mm |
€ 85.65
Pack of 50 |
Estimated Shipment: 17-06-2025
Log in to see stock. |
|||||
10776665
|
Poeppelmann™
32000500000 |
50 mm | 22 mm |
€ 67.45
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10287254
|
Poeppelmann™
32000550000 |
55 mm | 22 mm |
€ 91.70
Pack of 50 |
Estimated Shipment: 03-06-2025
Log in to see stock. |
|||||
10575674
|
Poeppelmann™
32000700000 |
70 mm | 22 mm |
€ 121.00
Pack of 50 |
Estimated Shipment: 14-05-2025
Log in to see stock. |
|||||
10737065
|
Poeppelmann™
32007030000 |
703 mm | 22 mm |
€ 99.80
Pack of 50 |
Estimated Shipment: 03-06-2025
Log in to see stock. |
|||||
10145634
|
Poeppelmann™
32000760000 |
76 mm | 22 mm |
€ 111.43
Pack of 25 |
Estimated Shipment: 26-06-2025
Log in to see stock. |
|||||
Description
- Solid design for increased demands
- Acc. with DIN EN 10220, DIN EN 10255 and DIN EN ISO 1127
- High visual appeal due to grainded top surface finish and chamfered edges
Specifications
Ribbed Closure Plug | |
DIN EN 10220, DIN EN 10255 and DIN EN ISO 1127th | |
For high loading capacity |
Product Title
Poeppelmann™ KAPSTO™ GPN 320 Finned Plugs