Learn More
Poeppelmann™ KAPSTO™ GPN 320 Finned Plugs
Featuring ribs on the shake to provide a secure fit even with varying tube tolerances. Poeppelmann™ KAPSTO™ GPN 320 Finned Plugs are manufactured from black PE-LLD plastic.
€ 20.55 - € 133.00
Specifications
Product Type Ribbed Closure Plug
Certifications/Compliance DIN EN 10220, DIN EN 10255 and DIN EN ISO 1127th
For Use With (Application) For high loading capacity
View More Specs
Products 13
Product Code Brand Diameter (Metric) Outer Height (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Outer Height (Metric) Price Quantity & Availability  
10615885
View Documents
Poeppelmann™
32001020000
102 mm 22 mm
€ 133.00
Pack of 25
Estimated Shipment: 12-05-2025
to see stock.
Add to basket
10044014
View Documents
Poeppelmann™
32000180000
18 mm 15 mm
€ 20.55
Pack of 100
Estimated Shipment: 21-05-2025
to see stock.
Add to basket
10298284
View Documents
Poeppelmann™
32000200000
20 mm 15 mm
€ 23.70
Pack of 100
Estimated Shipment: 23-06-2025
to see stock.
Add to basket
10707605
View Documents
Poeppelmann™
32000320000
32 mm 19 mm
€ 58.55
Pack of 100
Estimated Shipment: 03-06-2025
to see stock.
Add to basket
10388654
View Documents
Poeppelmann™
32000340000
34 mm 19 mm
€ 65.25
Pack of 100
Estimated Shipment: 03-06-2025
to see stock.
Add to basket
10398654
View Documents
Poeppelmann™
32000380000
38 mm 19 mm
€ 82.80
Pack of 100
Estimated Shipment: 03-06-2025
to see stock.
Add to basket
10490045
View Documents
Poeppelmann™
32000400000
40 mm 19 mm
€ 103.00
Pack of 100
Estimated Shipment: 01-07-2025
to see stock.
Add to basket
10748175
View Documents
Poeppelmann™
32000480000
48 mm 19 mm
€ 85.65
Pack of 50
Estimated Shipment: 17-06-2025
to see stock.
Add to basket
10776665
View Documents
Poeppelmann™
32000500000
50 mm 22 mm
€ 67.45
Pack of 50
10287254
View Documents
Poeppelmann™
32000550000
55 mm 22 mm
€ 91.70
Pack of 50
Estimated Shipment: 03-06-2025
to see stock.
Add to basket
10575674
View Documents
Poeppelmann™
32000700000
70 mm 22 mm
€ 121.00
Pack of 50
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
10737065
View Documents
Poeppelmann™
32007030000
703 mm 22 mm
€ 99.80
Pack of 50
Estimated Shipment: 03-06-2025
to see stock.
Add to basket
10145634
View Documents
Poeppelmann™
32000760000
76 mm 22 mm
€ 111.43
Pack of 25
Estimated Shipment: 26-06-2025
to see stock.
Add to basket
Description
Description
  • Solid design for increased demands
  • Acc. with DIN EN 10220, DIN EN 10255 and DIN EN ISO 1127
  • High visual appeal due to grainded top surface finish and chamfered edges
Specifications
Specifications
Ribbed Closure Plug
DIN EN 10220, DIN EN 10255 and DIN EN ISO 1127th
For high loading capacity
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Poeppelmann™ KAPSTO™ GPN 320 Finned Plugs