Learn More
Poeppelmann™ KAPSTO™ GPN 320 Finned Plugs
Featuring ribs on the shake to provide a secure fit even with varying tube tolerances. Poeppelmann™ KAPSTO™ GPN 320 Finned Plugs are manufactured from black PE-LLD plastic.
Brand: Poeppelmann™ 32000480000
This item is not returnable.
View return policy
Description
- Solid design for increased demands
- Acc. with DIN EN 10220, DIN EN 10255 and DIN EN ISO 1127
- High visual appeal due to grainded top surface finish and chamfered edges
Specifications
DIN EN 10220, DIN EN 10255 and DIN EN ISO 1127th | |
Ribbed Closure Plug | |
19 mm |
48 mm | |
For high loading capacity |
Product Content Correction
Product Title
Poeppelmann™ KAPSTO™ GPN 320 Finned Plugs > Diameter: 48 mm; Height: 19 mm