Learn More
Poeppelmann™ KAPSTO™ GPN 320 Finned Plugs
Featuring ribs on the shake to provide a secure fit even with varying tube tolerances. Poeppelmann™ KAPSTO™ GPN 320 Finned Plugs are manufactured from black PE-LLD plastic.
Brand:  Poeppelmann™ 32000400000
 View more versions of this product
Product Code. 10490045
  • € 97.50 / Pack of 100
Estimated Shipment: 18-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Solid design for increased demands
  • Acc. with DIN EN 10220, DIN EN 10255 and DIN EN ISO 1127
  • High visual appeal due to grainded top surface finish and chamfered edges
Specifications
Specifications
DIN EN 10220, DIN EN 10255 and DIN EN ISO 1127th
Ribbed Closure Plug
19 mm
40 mm
For high loading capacity
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers