Learn More
Poeppelmann™ KAPSTO™ GPN 320 Finned Plugs
Product Code. 10398654
Click to view available options
Diameter (Metric) Outer:
18 mm
20 mm
32 mm
34 mm
38 mm
40 mm
48 mm
50 mm
55 mm
70 mm
76 mm
102 mm
703 mm
Height (Metric):
15 mm
19 mm
22 mm
Unit Size:
Pack of 100
Pack of 25
Pack of 50
Product Code. 10398654
Brand: Poeppelmann™ 32000380000
Only null left
Add to basket
Add to basket
Featuring ribs on the shake to provide a secure fit even with varying tube tolerances. Poeppelmann™ KAPSTO™ GPN 320 Finned Plugs are manufactured from black PE-LLD plastic.
  • Solid design for increased demands
  • Acc. with DIN EN 10220, DIN EN 10255 and DIN EN ISO 1127
  • High visual appeal due to grainded top surface finish and chamfered edges
Specifications
Certifications/Compliance DIN EN 10220, DIN EN 10255 and DIN EN ISO 1127th
Diameter (Metric) Outer 38 mm
Product Type Ribbed Closure Plug
For Use With (Application) For high loading capacity
Height (Metric) 19 mm