Learn More
Alpha Packaging™ Clear Laboratory Glassware Pot
Use the Alpha Packaging™ Clear Laboratory Glassware Pot as a fundamental piece of laboratory glassware. The autoclavable clear glass is breakproof and chemically and thermally shockproof.
€ 13.95 - € 108.00
Product Code | Brand | Capacity (Metric) | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Quantity | Price | Quantity & Availability | ||||||
10554375
|
Alpha Packaging™
1000732 |
1000 mL | 18 Set |
€ 104.00
Pack of 18 |
Estimated Shipment: 10-06-2025
Log in to see stock. |
||||||
10647075
|
Alpha Packaging™
1000729 |
100 mL | 63 Set |
€ 84.10
Pack of 63 |
In Stock
Log in to see stock. |
||||||
10744347
|
Alpha Packaging™
231265 |
150 mL | 42 Set |
€ 81.20
Pack of 42 |
In Stock
Log in to see stock. |
||||||
10310015
|
Alpha Packaging™
601115 |
370 mL | 12 Set | N/A | N/A | N/A | |||||
10195884
|
Alpha Packaging™
1000721 |
250 mL | 48 Set |
€ 108.00
Pack of 48 |
In Stock
Log in to see stock. |
||||||
10647845
|
Alpha Packaging™
231263 |
100 mL | 72 Set | N/A | N/A | N/A | |||||
10207994
|
Alpha Packaging™
1000716 |
500 mL | 20 Set |
€ 50.15
Pack of 20 |
In Stock
Log in to see stock. |
||||||
Description
- Clear borosilicate glass
- Break-resistant
- Chemically and thermally shockproof;
- In compliance with EP, USP and JP guidelines
- Autoclavable
Specifications
Glass | |
Pot |
Product Title
Alpha Packaging™ Clear Laboratory Glassware Pot