Learn More
Alpha Packaging™ Clear Laboratory Glassware Pot
Use the Alpha Packaging™ Clear Laboratory Glassware Pot as a fundamental piece of laboratory glassware. The autoclavable clear glass is breakproof and chemically and thermally shockproof.
Brand:  Alpha Packaging™ 231263
 View more versions of this product
Product Code. 10647845
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Clear borosilicate glass
  • Break-resistant
  • Chemically and thermally shockproof;
  • In compliance with EP, USP and JP guidelines
  • Autoclavable
Specifications
Specifications
DIN 45
100 mL
Pot
72 Set
Glass