Learn More
Alpha Packaging™ Clear Laboratory Glassware Pot
Use the Alpha Packaging™ Clear Laboratory Glassware Pot as a fundamental piece of laboratory glassware. The autoclavable clear glass is breakproof and chemically and thermally shockproof.
Brand:  Alpha Packaging™ 231271
 View more versions of this product
Product Code. 10207994
  • € 50.15 / Pack of 20
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Clear borosilicate glass
  • Break-resistant
  • Chemically and thermally shockproof;
  • In compliance with EP, USP and JP guidelines
  • Autoclavable
Specifications
Specifications
DIN 55
500 mL
Pot
20 Set
Glass