Learn More
Alpha Packaging™ Clear Laboratory Glassware Pot
Use the Alpha Packaging™ Clear Laboratory Glassware Pot as a fundamental piece of laboratory glassware. The autoclavable clear glass is breakproof and chemically and thermally shockproof.
Brand:  Alpha Packaging™ 1000729
 View more versions of this product
Product Code. 10647075
  • € 84.10 / Pack of 63
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Clear borosilicate glass
  • Break-resistant
  • Chemically and thermally shockproof;
  • In compliance with EP, USP and JP guidelines
  • Autoclavable
Specifications
Specifications
DIN 40
100 mL
Pot
63 Set
Glass
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Alpha Packaging™ Clear Laboratory Glassware Pot > Capacity: 100mL; Quantity: 63 Set