Learn More
Berlin Packaging™ Clear Laboratory Glassware Pot
Product Code. 10554375
Click to view available options
Capacity (Metric):
100 mL
150 mL
250 mL
370 mL
500 mL
1000 mL
Quantity:
12 Set
18 Set
20 Set
42 Set
48 Set
63 Set
72 Set
Unit Size:
Pack of 12
Pack of 18
Pack of 20
Pack of 2688
Pack of 4158
Pack of 42
Pack of 48
Pack of 63
Pack of 72
Pack of 720
Product Code. 10554375
Brand: Berlin Packaging™ 1000732
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use the Berlin Packaging™ Clear Laboratory Glassware Pot as a fundamental piece of laboratory glassware. The autoclavable clear glass is breakproof and chemically and thermally shockproof.
  • Clear borosilicate glass
  • Break-resistant
  • Chemically and thermally shockproof;
  • In compliance with EP, USP and JP guidelines
  • Autoclavable
Specifications
Certifications/Compliance DIN 68
Quantity 18 Set
Capacity (Metric) 1000 mL
Material Glass
Product Type Pot