Learn More
Alpha Packaging™ Clear Laboratory Glassware Pot
Use the Alpha Packaging™ Clear Laboratory Glassware Pot as a fundamental piece of laboratory glassware. The autoclavable clear glass is breakproof and chemically and thermally shockproof.
Brand: Alpha Packaging™ 601115
This item is not returnable.
View return policy
Description
- Clear borosilicate glass
- Break-resistant
- Chemically and thermally shockproof;
- In compliance with EP, USP and JP guidelines
- Autoclavable
Specifications
DIN 63 | |
370 mL | |
Pot |
12 Set | |
Glass |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Alpha Packaging™ Clear Laboratory Glassware Pot > Capacity: 370mL; Quantity: 12 Set