Learn More
MK-2206 dihydrochloride, MedChemExpress
MedChemExpress MK-2206 dihydrochloride (MK-2206 (2HCl)) is an orally active allosteric AKT inhibitor with IC50s of 5 nM, 12 nM, and 65 nM for AKT1, AKT2, and AKT3, respectively. MK-2206 dihydrochloride induces autophagy.
Brand:  Medchem Express HY-10358
Encompass_Preferred
 View more versions of this product
Product Code. 16450237
  • € 291.60 / 50mg
Explore more special offers
Description
Description
MK-2206 dihydrochloride (MK-2206 (2HCl)) is an orally active allosteric AKT inhibitor with IC50s of 5 nM, 12 nM, and 65 nM for AKT1, AKT2, and AKT3, respectively. MK-2206 dihydrochloride induces autophagy.
Specifications
Specifications
MK-2206 dihydrochloride
Green Yellow
C25H23Cl2N5O
MK-2206 (2HCl)
O=C1N2C(C3=CC(C4=CC=CC=C4)=C(N=C3C=C2)C5=CC=C(C6(N)CCC6)C=C5)=NN1.Cl.Cl
480.39
Research
Cancer-Kinase/protease
1032350-13-2
Research
50 mg
DMSO : 16.67 mg/mL (34.70 mM; ultrasonic and warming and heat to 70°C) ∣H2O : 3.57 mg/mL (7.43 mM; ultrasonic and warming and heat to 60°C)
480.39
98.89%
Solid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
MK-2206 dihydrochloride, MedChemExpress
Research purposes only