Learn More
MK-2206 dihydrochloride, MedChemExpress

MedChemExpress MK-2206 dihydrochloride (MK-2206 (2HCl)) is an orally active allosteric AKT inhibitor with IC50s of 5 nM, 12 nM, and 65 nM for AKT1, AKT2, and AKT3, respectively. MK-2206 dihydrochloride induces autophagy.

€ 93.15 - € 754.50
Specifications
Chemical Name or Material | MK-2206 dihydrochloride |
---|---|
CAS | 1032350-13-2 |
Color | Green Yellow |
Purity Grade Notes | Research |
Molecular Formula | C25H23Cl2N5O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16496587
![]() |
Medchem Express
HY-10358 |
5 mg |
€ 93.15
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16460976
![]() |
Medchem Express
HY-10358 |
200 mg |
€ 754.50
200mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16450237
![]() |
Medchem Express
HY-10358 |
50 mg |
€ 291.60
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16423868
![]() |
Medchem Express
HY-10358 |
10 mg |
€ 136.50
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16454896
![]() |
Medchem Express
HY-10358 |
100 mg |
€ 483.00
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MK-2206 dihydrochloride (MK-2206 (2HCl)) is an orally active allosteric AKT inhibitor with IC50s of 5 nM, 12 nM, and 65 nM for AKT1, AKT2, and AKT3, respectively. MK-2206 dihydrochloride induces autophagy.Specifications
MK-2206 dihydrochloride | |
Green Yellow | |
C25H23Cl2N5O | |
DMSO : 16.67 mg/mL (34.70 mM; ultrasonic and warming and heat to 70°C) ∣H2O : 3.57 mg/mL (7.43 mM; ultrasonic and warming and heat to 60°C) | |
480.39 | |
98.89% | |
Solid |
1032350-13-2 | |
Research | |
MK-2206 (2HCl) | |
O=C1N2C(C3=CC(C4=CC=CC=C4)=C(N=C3C=C2)C5=CC=C(C6(N)CCC6)C=C5)=NN1.Cl.Cl | |
480.39 | |
Research | |
Cancer-Kinase/protease |
Safety and Handling
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.