Learn More
Afuresertib hydrochloride, MedChemExpress
MedChemExpress Afuresertib hydrochloride (GSK 2110183 hydrochloride) is an orally bioavailable, selective, ATP-competitive and potent pan-Akt kinase inhibitor with Kis of 0.08/2/2.6 nM for Akt1/Akt2/Akt3 respectively.
Brand:  Medchem Express HY-15727A
Encompass_Preferred
 View more versions of this product
Product Code. 16412857
  • € 498.15 / 50mg
Explore more special offers
Description
Description
Afuresertib hydrochloride (GSK 2110183 hydrochloride) is an orally bioavailable, selective, ATP-competitive and potent pan-Akt kinase inhibitor with Kis of 0.08/2/2.6 nM for Akt1/Akt2/Akt3 respectively.
Specifications
Specifications
Afuresertib hydrochloride
White
C18H18Cl3FN4OS
GSK2110183 hydrochloride
O=C(C1=CC(C2=C(Cl)C=NN2C)=C(Cl)S1)N[C@@H](CC3=CC=CC(F)=C3)CN.[H]Cl
463.78
Research
Cancer-Kinase/protease
1047645-82-8
Research
50 mg
DMSO : 100 mg/mL (215.62 mM; Need ultrasonic)
463.78
98.08%
Solid
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Research purposes only