Learn More
Afuresertib hydrochloride, MedChemExpress
MedChemExpress Afuresertib hydrochloride (GSK 2110183 hydrochloride) is an orally bioavailable, selective, ATP-competitive and potent pan-Akt kinase inhibitor with Kis of 0.08/2/2.6 nM for Akt1/Akt2/Akt3 respectively.
Brand: Medchem Express HY-15727A
This item is not returnable.
View return policy
Description
Afuresertib hydrochloride (GSK 2110183 hydrochloride) is an orally bioavailable, selective, ATP-competitive and potent pan-Akt kinase inhibitor with Kis of 0.08/2/2.6 nM for Akt1/Akt2/Akt3 respectively.Specifications
Afuresertib hydrochloride | |
White | |
C18H18Cl3FN4OS | |
GSK2110183 hydrochloride | |
O=C(C1=CC(C2=C(Cl)C=NN2C)=C(Cl)S1)N[C@@H](CC3=CC=CC(F)=C3)CN.[H]Cl | |
463.78 | |
Research | |
Cancer-Kinase/protease |
1047645-82-8 | |
Research | |
5 mg | |
DMSO : 100 mg/mL (215.62 mM; Need ultrasonic) | |
463.78 | |
98.08% | |
Solid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Research purposes only