Learn More
Maprotiline hydrochloride, MedChemExpress
MedChemExpress Maprotiline hydrochloride is a selective noradrenalin re-uptake inhibitor and a tetracyclic antidepressant.
Brand:  Medchem Express HY-B0444
Encompass
 View more versions of this product
Product Code. 16428316
  • € 112.50 / 100mg
Explore more special offers
Description
Description
Maprotiline hydrochloride is a selective noradrenalin re-uptake inhibitor and a tetracyclic antidepressant.
Specifications
Specifications
Maprotiline hydrochloride
White
C20H24ClN
DMSO : 83.33 mg/mL (265.50 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 2.2 mg/mL (7.01 mM; Need ultrasonic)
313.86
99.43%
Solid
10347-81-6
Research
100 mg
CNCCCC1(CC2)C3=CC=CC=C3C2C4=CC=CC=C14.Cl
313.86
Research
Neuroscience-Neuromodulation
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Maprotiline hydrochloride, MedChemExpress
Research purposes only