Learn More
Maprotiline hydrochloride, MedChemExpress

MedChemExpress Maprotiline hydrochloride is a selective noradrenalin re-uptake inhibitor and a tetracyclic antidepressant.

€ 112.50 - € 195.75
Specifications
Chemical Name or Material | Maprotiline hydrochloride |
---|---|
CAS | 10347-81-6 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C20H24ClN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16492136
![]() |
Medchem Express
HY-B0444 |
500 mg |
€ 195.75
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16428316
![]() |
Medchem Express
HY-B0444 |
100 mg |
€ 112.50
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Maprotiline hydrochloride is a selective noradrenalin re-uptake inhibitor and a tetracyclic antidepressant.Specifications
Maprotiline hydrochloride | |
White | |
C20H24ClN | |
CNCCCC1(CC2)C3=CC=CC=C3C2C4=CC=CC=C14.Cl | |
313.86 | |
Research | |
Neuroscience-Neuromodulation |
10347-81-6 | |
Research | |
DMSO : 83.33 mg/mL (265.50 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 2.2 mg/mL (7.01 mM; Need ultrasonic) | |
313.86 | |
99.43% | |
Solid |
Safety and Handling
H302
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.