missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Cylindrical Funnel with Sintered Glass Disc
Available with different porosity (grades) glass sintered discs for the filtration of a range of precipitates
€ 26.10 - € 605.00
Product Code | Brand | Capacity (Metric) | Pore Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Pore Size | Price | Quantity & Availability | ||||
15577720
|
Pyrex
3770/10 |
50 mL | 5 |
€ 229.00
Pack of 10 |
Estimated Shipment: 03-01-2025
Log in to see stock. |
||||
15547720
|
Pyrex
3770/04 |
50 mL | 40 to 100μm |
€ 605.00
Pack of 10 |
Estimated Shipment: 03-01-2025
Log in to see stock. |
||||
15567720
|
Pyrex
3770/08 |
50 mL | 4 |
€ 449.00
Pack of 10 |
Estimated Shipment: 17-01-2025
Log in to see stock. |
||||
15537720
|
Pyrex
3770/02 |
50 mL | 100 to 160μm |
€ 605.00
Pack of 10 |
Estimated Shipment: 17-01-2025
Log in to see stock. |
||||
15557720
|
Pyrex
3770/06 |
50 mL | 3 |
€ 230.00
Pack of 10 |
Estimated Shipment: 17-01-2025
Log in to see stock. |
||||
15527720
|
Pyrex
3760/04 |
130 mL | 2 | ||||||
15517720
|
Pyrex
3760/02 |
130 mL | 1 | ||||||
11348014
|
Pyrex
3760/06 |
130 mL | 3 | ||||||
Description
- Manufactured from Pyrex™ borosilicate glass
- Ideal for filtering precipitates
- 3770 series 20mm diameter sintered discs
- 3760 series 30mm diameter sintered discs