Learn More
Pyrex™ Cylindrical Funnel with Sintered Glass Disc
Available with different porosity (grades) glass sintered discs for the filtration of a range of precipitates
Brand:  Pyrex 3770/10
217.55 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 15577720
  • € 229.00 / Pack of 10
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Description
  • Manufactured from Pyrex™ borosilicate glass
  • Ideal for filtering precipitates
  • 3770 series 20mm diameter sintered discs
  • 3760 series 30mm diameter sintered discs
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50 mL
Borosilicate Glass
160 mm
5