Learn More
Pyrex™ Cylindrical Funnel with Sintered Glass Disc
Available with different porosity (grades) glass sintered discs for the filtration of a range of precipitates
Brand:  Pyrex 3760/04
 View more versions of this product
Product Code. 15527720
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Manufactured from Pyrex™ borosilicate glass
  • Ideal for filtering precipitates
  • 3770 series 20mm diameter sintered discs
  • 3760 series 30mm diameter sintered discs
TRUSTED_SUSTAINABILITY
Specifications
Specifications
130 mL
Pyrex Glass, Borosilicate Glass
For filtering precipitates
Sintered glass disc
Filter funnel
300 mm
30mm
2