missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Cylindrical Funnel with Sintered Glass Disc
Available with different porosity (grades) glass sintered discs for the filtration of a range of precipitates
Brand: Pyrex 3770/02
574.75 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Manufactured from Pyrex™ borosilicate glass
- Ideal for filtering precipitates
- 3770 series 20mm diameter sintered discs
- 3760 series 30mm diameter sintered discs
Specifications
50 mL | |
Borosilicate Glass | |
160 mm | |
100 to 160μm |