Learn More
Poeppelmann™ KAPSTO™ GPN 670 Flange Covers
Closes the opening and covers the sealing face and internal threads while preventing accidental dislodging. Poeppelmann™ KAPSTO™ GPN 670 Flange Covers are manufactured from yellow PE-LLD/PE-HD plastic.
€ 47.20 - € 403.00
Product Code | Brand | Diameter (Metric) Outside | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outside | Price | Quantity & Availability | |||||
10073934
|
Poeppelmann™
67000650000 |
180 mm |
€ 97.50
Pack of 50 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
10686245
|
Poeppelmann™
67002500000 |
400 mm |
€ 403.00
Pack of 25 |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||||
10482565
|
Poeppelmann™
67000200000 |
100 mm |
€ 53.95
Pack of 100 |
Estimated Shipment: 04-02-2025
Log in to see stock. |
|||||
10175174
|
Poeppelmann™
67000250000 |
110 mm |
€ 91.15
Pack of 100 |
Estimated Shipment: 04-02-2025
Log in to see stock. |
|||||
10747225
|
Poeppelmann™
67000500000 |
140 mm |
€ 169.00
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10708595
|
Poeppelmann™
67000150000 |
90 mm |
€ 47.20
Pack of 100 |
Estimated Shipment: 13-02-2025
Log in to see stock. |
|||||
10470835
|
Poeppelmann™
67002000000 |
345 mm |
€ 284.00
Pack of 25 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
10319024
|
Poeppelmann™
67000400000 |
146 mm |
€ 142.00
Pack of 100 |
Estimated Shipment: 04-02-2025
Log in to see stock. |
|||||
Description
- Acc. with DIN EN 12351
- Larger tolerances accomodated by ribs
- Suitable for PN 10 through PN 400 and DN 10 through DN 250