Learn More
Poeppelmann™ KAPSTO™ GPN 670 Flange Covers
Closes the opening and covers the sealing face and internal threads while preventing accidental dislodging. Poeppelmann™ KAPSTO™ GPN 670 Flange Covers are manufactured from yellow PE-LLD/PE-HD plastic.
€ 46.05 - € 383.00
Product Code | Brand | Diameter (Metric) Outside | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outside | Price | Quantity & Availability | |||||
10073934
|
Poeppelmann™
67000650000 |
180 mm |
€ 92.75
Pack of 50 |
Estimated Shipment: 22-10-2024
Log in to see stock. |
|||||
10686245
|
Poeppelmann™
67002500000 |
400 mm |
€ 383.00
Pack of 25 |
Estimated Shipment: 04-10-2024
Log in to see stock. |
|||||
10482565
|
Poeppelmann™
67000200000 |
100 mm |
€ 51.30
Pack of 100 |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10175174
|
Poeppelmann™
67000250000 |
110 mm |
€ 86.70
Pack of 100 |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10747225
|
Poeppelmann™
67000500000 |
140 mm |
€ 160.00
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10708595
|
Poeppelmann™
67000150000 |
90 mm |
€ 46.05
Pack of 100 |
Estimated Shipment: 23-10-2024
Log in to see stock. |
|||||
10470835
|
Poeppelmann™
67002000000 |
345 mm |
€ 270.10
Pack of 25 |
Estimated Shipment: 22-10-2024
Log in to see stock. |
|||||
10319024
|
Poeppelmann™
67000400000 |
146 mm |
€ 137.10
Pack of 100 |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
Description
- Acc. with DIN EN 12351
- Larger tolerances accomodated by ribs
- Suitable for PN 10 through PN 400 and DN 10 through DN 250