Learn More
Poeppelmann™ KAPSTO™ GPN 670 Flange Covers
Closes the opening and covers the sealing face and internal threads while preventing accidental dislodging. Poeppelmann™ KAPSTO™ GPN 670 Flange Covers are manufactured from yellow PE-LLD/PE-HD plastic.
Brand: Poeppelmann™ 67000250000
This item is not returnable.
View return policy
Description
- Acc. with DIN EN 12351
- Larger tolerances accomodated by ribs
- Suitable for PN 10 through PN 400 and DN 10 through DN 250
Specifications
Yellow | |
Suitable for BASF-Werknorm 14-150. and screw holes | |
110 mm | |
Flange Cover Plug |
Closes the opening and covers sealing face and internal threads | |
100 Pack | |
LDPE |