Learn More
Poeppelmann™ KAPSTO™ GPN 670 Flange Covers
Closes the opening and covers the sealing face and internal threads while preventing accidental dislodging. Poeppelmann™ KAPSTO™ GPN 670 Flange Covers are manufactured from yellow PE-LLD/PE-HD plastic.
Brand:  Poeppelmann™ 67000200000
 View more versions of this product
Product Code. 10482565
  • € 53.95 / Pack of 100
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Acc. with DIN EN 12351
  • Larger tolerances accomodated by ribs
  • Suitable for PN 10 through PN 400 and DN 10 through DN 250
Specifications
Specifications
Yellow
Suitable for BASF-Werknorm 14-150. and screw holes
100 mm
Flange Cover Plug
Closes the opening and covers sealing face and internal threads
100 Pack
LDPE
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers