Learn More
Poeppelmann™ KAPSTO™ GPN 670 Flange Covers
Closes the opening and covers the sealing face and internal threads while preventing accidental dislodging. Poeppelmann™ KAPSTO™ GPN 670 Flange Covers are manufactured from yellow PE-LLD/PE-HD plastic.
Brand:  Poeppelmann™ 67000150000
 View more versions of this product
Product Code. 10708595
  • € 46.05 / Pack of 100
Estimated Shipment: 23-10-2024
to see stock.
Explore more special offers
Description
  • Acc. with DIN EN 12351
  • Larger tolerances accomodated by ribs
  • Suitable for PN 10 through PN 400 and DN 10 through DN 250
Specifications
Specifications
Yellow
100/ck.
LDPE
Closes the opening and covers sealing face and internal threads
90 mm
Flange Cover Plug