Learn More
Product Code | Brand | Diameter (Metric) Outer | Height (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Height (Metric) | Price | Quantity & Availability | |||||
10460255
|
Poeppelmann™
61002910000 |
45 mm | 13 mm |
€ 38.35
Pack of 100 |
Estimated Shipment: 06-05-2025
Log in to see stock. |
|||||
10697215
|
Poeppelmann™
61002410000 |
33 mm | 19 mm |
€ 38.69
Pack of 100 |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
10696455
|
Poeppelmann™
61000310000 |
48 mm | 19 mm |
€ 46.65
Pack of 100 |
Estimated Shipment: 09-05-2025
Log in to see stock. |
|||||
10491605
|
Poeppelmann™
61000420000 |
83 mm | 19 mm |
€ 111.24
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10329164
|
Poeppelmann™
61000430000 |
92 mm | 19 mm |
€ 117.42
Pack of 100 |
In Stock
Log in to see stock. ![]() |
|||||
10481415
|
Poeppelmann™
61000400000 |
71 mm | 29 mm |
€ 94.31
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10481025
|
Poeppelmann™
61000360000 |
62 mm | 25 mm |
€ 80.74
Pack of 100 |
Estimated Shipment: 01-05-2025
Log in to see stock. |
|||||
10420265
|
Poeppelmann™
61000340000 |
54 mm | 19 mm |
€ 56.26
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10105314
|
Poeppelmann™
61000190000 |
27 mm | 16 mm |
€ 17.58
Pack of 100 |
In Stock
Log in to see stock. ![]() |
|||||
Description
- Universal use as a plug or cap
- Acc. with VDA Band 19 and ISO 16232
- Tapered design ideal for accomodating tolerances
Product Title
Poeppelmann™ KAPSTO™ GPN 610 Universal Protection Caps/Plugs