Learn More
Poeppelmann™ KAPSTO™ GPN 610 Universal Protection Caps/Plugs
Excellent for closing off bore holes and internal threads, or to protect studs and external threads. Poeppelmann™ KAPSTO™ GPN 610 Universal Protection Caps/Plugs are manufactured from yellow PE-LD plastic.
Brand:  Poeppelmann™ 61000310000
 View more versions of this product
Product Code. 10696455
  • € 46.65 / Pack of 100
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Universal use as a plug or cap
  • Acc. with VDA Band 19 and ISO 16232
  • Tapered design ideal for accomodating tolerances
Specifications
Specifications
48 mm
Universal Protection Closure
For sealing holes and internal threads or to protect bolts and outer threads
19 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Poeppelmann™ KAPSTO™ GPN 610 Universal Protection Caps/Plugs > Size: 19 H x 41.3 I.D. x 48mm O.D.