Learn More
Lenz Laborglasintrumente Flat-Bottom Flasks With Conical Ground Joint
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
€ 75.00 - € 825.00
Specifications
Includes Conical ground joint and inscription label
View More Specs
Products 9
Product Code Brand Capacity (Metric) Standard Taper Joint Diameter (Metric) Outer Price Quantity & Availability  
Product Code Brand Capacity (Metric) Standard Taper Joint Diameter (Metric) Outer Price Quantity & Availability  
10141471
View Documents
Lenz Laborglasintrumente
03011937
100 mL NS 19/26 64 mm
€ 157.00
Pack of 10
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
10643501
View Documents
Lenz Laborglasintrumente
03012937
100 mL NS 29/32 64 mm
€ 75.00
Pack of 10
Estimated Shipment: 01-05-2025
to see stock.
Add to basket
10433491
View Documents
Lenz Laborglasintrumente
03012973
2 L NS 29/32 166 mm
€ 727.00
Pack of 10
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
10060211
View Documents
Lenz Laborglasintrumente
03011949
250 mL NS 19/26 85 mm
€ 274.00
Pack of 10
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
10274141
View Documents
Lenz Laborglasintrumente
03012473
2 L NS 24/29 166 mm
€ 825.00
Pack of 10
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
10460703
View Documents
Lenz Laborglasintrumente
03012428
50 mL NS 24/29 51 mm
€ 130.00
Pack of 10
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
10500122
View Documents
Lenz Laborglasintrumente
03012437
100 mL NS 24/29 64 mm
€ 149.00
Pack of 10
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
10694382
View Documents
Lenz Laborglasintrumente
03011958
500 mL NS 19/26 105 mm
€ 406.00
Pack of 10
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
10767891
View Documents
Lenz Laborglasintrumente
03012470
1 L NS 24/29 131 mm
€ 502.00
Pack of 10
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
Description
Description
  • According to DIN EN ISO 4797
  • Wiith inscription label
  • Material: DURAN™
Specifications
Specifications
Conical ground joint and inscription label
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Lenz Laborglasintrumente Flat-Bottom Flasks With Conical Ground Joint