Learn More
Lenz Laborglasintrumente Flat-Bottom Flasks With Conical Ground Joint
Product Code. 10460703
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1 L
2 L
Standard Taper Joint:
NS 19/26
NS 24/29
NS 29/32
Diameter (Metric) Outer:
51 mm
64 mm
85 mm
105 mm
131 mm
166 mm
Unit Size:
Pack of 10
Product Code. 10460703
Brand: Lenz Laborglasintrumente 03012428
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • According to DIN EN ISO 4797
  • Wiith inscription label
  • Material: DURAN™
Specifications
Capacity (Metric) 50 mL
Includes Conical ground joint and inscription label
Standard Taper Joint NS 24/29
Diameter (Metric) Outer 51 mm
Quantity 10/Pk.