Learn More
B Braun™ Vasco™OP Sensitive Sterile Glove
Gain a better grip with micro-rough surface glove. B Braun™ Vasco™OP Sensitive Sterile Glove with inner coating is made of highly elastic natural latex. These ergonomically shaped surgical gloves are comfortable, provide good tactile sensation and feature rolled cuffs.  
€ 2.00 - € 69.00
Specifications
Cut Style Rolled cuff
Color Tan
Latex Free No
Material Latex
Powdered Powder Free
View More Specs
Alert:
Warning: Gloves contain natural latex, the allergic Reactions including anaphylactic reactions can trigger
Products 5
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
10094940
View Documents
B Braun™
6081002
6 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10527604
View Documents
B Braun™
6081037
7.5
€ 62.00
Pack of 40
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
10461761
View Documents
B Braun™
6081037
7.5
€ 2.00
1 pair
In Stock
to see stock.
Edge
Add to basket
10412365
View Documents
B Braun™
6081045
8
€ 69.00
Pack of 40
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
10219080
View Documents
B Braun™
6081045
8 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Medical surgical glove according to EN 455
  • Protective gloves according to PSA 89/686 / EEC, EN 420, EN 374
  • Easier working due to fully anatomical shape
  • Very good grip thanks to micro-rough surface
  • Excellent wearing comfort and excellent fit
  • Perfect tactile sensitivity and optimal safety
  • AQL: 0.65
Specifications
Specifications
Rolled cuff
No
Powder Free
Tan
Tan
Latex