Learn More
B Braun™ Vasco™OP Sensitive Sterile Glove
Gain a better grip with micro-rough surface glove. B Braun™ Vasco™OP Sensitive Sterile Glove with inner coating is made of highly elastic natural latex. These ergonomically shaped surgical gloves are comfortable, provide good tactile sensation and feature rolled cuffs.
Brand: B Braun™ 6081002
Alert:
Warning: Gloves contain natural latex, the allergic Reactions including anaphylactic reactions can trigger
Description
- Medical surgical glove according to EN 455
- Protective gloves according to PSA 89/686 / EEC, EN 420, EN 374
- Easier working due to fully anatomical shape
- Very good grip thanks to micro-rough surface
- Excellent wearing comfort and excellent fit
- Perfect tactile sensitivity and optimal safety
- AQL: 0.65
Specifications
Rolled cuff | |
No | |
Powder Free | |
Sterile |
Tan | |
Latex | |
6 | |
Tan |