Learn More
B Braun™ Vasco™OP Sensitive Sterile Glove
Gain a better grip with micro-rough surface glove. B Braun™ Vasco™OP Sensitive Sterile Glove with inner coating is made of highly elastic natural latex. These ergonomically shaped surgical gloves are comfortable, provide good tactile sensation and feature rolled cuffs.  
Brand:  B Braun™ 6081045
 View more versions of this product
Product Code. 10412365
  • € 69.00 / Pack of 40
Estimated Shipment: 05-12-2024
to see stock.
Explore more special offers
Add to basket
Alert:
Warning: Gloves contain natural latex, the allergic Reactions including anaphylactic reactions can trigger
Description
Description
  • Medical surgical glove according to EN 455
  • Protective gloves according to PSA 89/686 / EEC, EN 420, EN 374
  • Easier working due to fully anatomical shape
  • Very good grip thanks to micro-rough surface
  • Excellent wearing comfort and excellent fit
  • Perfect tactile sensitivity and optimal safety
  • AQL: 0.65
Specifications
Specifications
Rolled cuff
No
Powder Free
Sterile
Tan
Latex
8
Tan
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers