Learn More
Thermo Scientific™ SUN-SRi™ MicroMat™ PTFE-Coated Silicone Sealing Mats
Provides superior resealability after up to 100 injections, resists coring and tearing
POA
Specifications
Autoclavable Autoclavable
No. of Wells 96
No. per Case 10
Product Type Sealing Mat
View More Specs
Products 1
Product Code Brand Color Material Well Shape Description Price Quantity & Availability  
Product Code Brand Color Material Well Shape Description Price Quantity & Availability  
12376962
Encompass_Preferred
View Documents
Thermo Scientific™
14-823-258
Blue Silicone, PTFE Square Flat Underside This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
  • PTFE-coated silicone
  • Eliminates cross contamination of samples
  • Available pre-slit for easy penetration and to reduce vacuum
  • Dry heat autoclavable
  • Superior chemical compatibility
Specifications
Specifications
Autoclavable
96
10
Sealing Mat