Learn More
Thermo Scientific™ SUN-SRi™ MicroMat™ PTFE-Coated Silicone Sealing Mats
Provides superior resealability after up to 100 injections, resists coring and tearing
Brand:  Thermo Scientific™ 14-823-258
Encompass_Preferred
 View more versions of this product
Product Code. 12376962
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • PTFE-coated silicone
  • Eliminates cross contamination of samples
  • Available pre-slit for easy penetration and to reduce vacuum
  • Dry heat autoclavable
  • Superior chemical compatibility
Specifications
Specifications
Blue
Silicone, PTFE
96
Yes
Webseal
Flat Underside
Autoclavable
10
Sealing Mat
Square