Learn More
Thermo Scientific™ SUN-SRi™ MicroMat™ PTFE-Coated Silicone Sealing Mats
Provides superior resealability after up to 100 injections, resists coring and tearing
POA
Specifications
Autoclavable | Autoclavable |
---|---|
No. of Wells | 96 |
No. per Case | 10 |
Product Type | Sealing Mat |
Product Code | Brand | Color | Material | Well Shape | Description | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Material | Well Shape | Description | Price | Quantity & Availability | ||
12376962
|
Thermo Scientific™
14-823-258 |
Blue | Silicone, PTFE | Square | Flat Underside | ||||
Description
- PTFE-coated silicone
- Eliminates cross contamination of samples
- Available pre-slit for easy penetration and to reduce vacuum
- Dry heat autoclavable
- Superior chemical compatibility
Specifications
Autoclavable | |
96 | |
10 | |
Sealing Mat |