Learn More
SP Bel-Art™ SP Bel-Art™ Sterileware Powder Funnels
Ideal for transferring powders or granules using a sterile sampling tool into a sterile container
€ 323.00 - € 444.00
Specifications
For Use With (Application) QC Powder Transfer
View More Specs
Products 4
Product Code Brand Diameter (Metric) Stem Capacity (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Stem Capacity (Metric) Price Quantity & Availability  
16815153
View Documents
SP Bel-Art™
F14660-1065
1.2 cm I.D. 49 mL
€ 323.00
Pack of 36
Estimated Shipment: 15-04-2025
to see stock.
Add to basket
16825153
View Documents
SP Bel-Art™
F14660-1080
1.5 cm I.D. 138 mL
€ 444.00
Pack of 36
Estimated Shipment: 15-04-2025
to see stock.
Add to basket
16835153
View Documents
SP Bel-Art™
F14660-1100
1.4 cm I.D. 248 mL
€ 323.00
Pack of 24
Estimated Shipment: 15-04-2025
to see stock.
Add to basket
16845153
View Documents
SP Bel-Art™
F14660-1150
2.4 cm I.D. 784 mL
€ 434.00
Pack of 24
Estimated Shipment: 17-02-2025
to see stock.
Add to basket
Description
Description
  • 60° angle and large bore stem help minimize bridging of powder
  • FDA grade polypropylene; safe for food, drug and cosmetic use
  • Gamma irradiated sterile (SAL 10^-6)
  • Individually bagged; ready for one time use and disposal
  • Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization
Specifications
Specifications
QC Powder Transfer