Learn More
SP Bel-Art™ SP Bel-Art™ Sterileware Powder Funnels
Ideal for transferring powders or granules using a sterile sampling tool into a sterile container
Brand:  SP Bel-Art™ F14660-1150
 View more versions of this product
Product Code. 16845153
  • € 434.00 / Pack of 24
Estimated Shipment: 28-02-2025
to see stock.
Explore more special offers
Description
  • 60° angle and large bore stem help minimize bridging of powder
  • FDA grade polypropylene; safe for food, drug and cosmetic use
  • Gamma irradiated sterile (SAL 10^-6)
  • Individually bagged; ready for one time use and disposal
  • Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization
Specifications
Specifications
784 mL
13.6 cm
3 cm
2.4 cm I.D.
QC Powder Transfer
24/Pk.