Learn More
SP Bel-Art™ SP Bel-Art™ Sterileware Powder Funnels
Ideal for transferring powders or granules using a sterile sampling tool into a sterile container
Brand: SP Bel-Art™ F14660-1100
This item is not returnable.
View return policy
Description
- 60° angle and large bore stem help minimize bridging of powder
- FDA grade polypropylene; safe for food, drug and cosmetic use
- Gamma irradiated sterile (SAL 10^-6)
- Individually bagged; ready for one time use and disposal
- Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization
Specifications
10 cm I.D. | |
1.4 cm I.D. | |
QC Powder Transfer | |
24/Pk. |
248 mL | |
8.3 cm | |
2.6 cm |
Product Content Correction
Product Title
SP Bel-Art™ SP Bel-Art™ Sterileware Powder Funnels > 1.4 cm I.D.