Learn More
Rebastinib, MedChemExpress

MedChemExpress Rebastinib (DCC-2036) is an orally active, non-ATP-competitive Bcr-Abl inhibitor for Abl1WT and Abl1T315I with IC50s of 0.8 nM and 4 nM, respectively. Rebastinib also inhibits SRC, KDR, FLT3, and Tie-2, and has low activity to seen towards c-Kit.

€ 117.45 - € 546.00
Specifications
Chemical Name or Material | Rebastinib |
---|---|
CAS | 1020172-07-9 |
Color | Off-White |
Purity Grade Notes | Research |
Molecular Formula | C30H28FN7O3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16461667
![]() |
Medchem Express
HY-13024 |
50 mg |
€ 546.00
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16448207
![]() |
Medchem Express
HY-13024 |
5 mg |
€ 117.45
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16405498
![]() |
Medchem Express
HY-13024 |
10 mg |
€ 195.75
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rebastinib (DCC-2036) is an orally active, non-ATP-competitive Bcr-Abl inhibitor for Abl1WT and Abl1T315I with IC50s of 0.8 nM and 4 nM, respectively. Rebastinib also inhibits SRC, KDR, FLT3, and Tie-2, and has low activity to seen towards c-Kit.Specifications
Rebastinib | |
Off-White | |
C30H28FN7O3 | |
DMSO : 50 mg/mL (90.32 mM; ultrasonic and warming and heat to 80°C) | |
553.59 | |
98.0% | |
Powder |
1020172-07-9 | |
Research | |
DCC-2036 | |
CC(C)(C)C1=NN(C(NC(NC2=C(F)C=C(OC3=CC(C(NC)=O)=NC=C3)C=C2)=O)=C1)C4=CC=C5C(C=CC=N5)=C4 | |
553.59 | |
Research | |
Cancer-Kinase/protease |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.