Learn More
Rebastinib, MedChemExpress
MedChemExpress Rebastinib (DCC-2036) is an orally active, non-ATP-competitive Bcr-Abl inhibitor for Abl1WT and Abl1T315I with IC50s of 0.8 nM and 4 nM, respectively. Rebastinib also inhibits SRC, KDR, FLT3, and Tie-2, and has low activity to seen towards c-Kit.
Brand:  Medchem Express HY-13024
Encompass_Preferred
 View more versions of this product
Product Code. 16405498
  • € 195.75 / 10mg
Explore more special offers
Description
Description
Rebastinib (DCC-2036) is an orally active, non-ATP-competitive Bcr-Abl inhibitor for Abl1WT and Abl1T315I with IC50s of 0.8 nM and 4 nM, respectively. Rebastinib also inhibits SRC, KDR, FLT3, and Tie-2, and has low activity to seen towards c-Kit.
Specifications
Specifications
Rebastinib
Off-White
C30H28FN7O3
DCC-2036
CC(C)(C)C1=NN(C(NC(NC2=C(F)C=C(OC3=CC(C(NC)=O)=NC=C3)C=C2)=O)=C1)C4=CC=C5C(C=CC=N5)=C4
553.59
Research
Cancer-Kinase/protease
1020172-07-9
Research
10 mg
DMSO : 50 mg/mL (90.32 mM; ultrasonic and warming and heat to 80°C)
553.59
98.0%
Powder
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Rebastinib, MedChemExpress
Research purposes only