Learn More
Lenz Laborglasinstrumente™ Pycnometers
Brand:  Lenz Laborglasinstrumente™ 05630213
 View more versions of this product
Product Code. 10375781
  • € 81.12 / Each
Estimated Shipment: 07-07-2025
to see stock.
Explore more special offers
Specifications
Pycnometer
10 mL
Pycnometer
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Pycnometers > Capacity: 10mL; Pear Shaped