Learn More
Cytiva ETFE Tubing
Brand:  Cytiva 18-1121-36
Product Code. 12534507
  • € 211.12 / 2m
In Stock
to see stock.
Explore more special offers
Specifications
Tubing
0.25 mm
2 m
ETFE Tubing, 2 m, i.d. 0.25 mm, o.d. 1/16 in.
1.58 mm
ETFE
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva ETFE Tubing > I.D.: 0.25mm